Product Details

SNP ID
rs1048503
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:228091322 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATAATGCCCTCCCTTAACTCAAG[A/G]CCAGCAGCAGATGTGTTAGAGGGGA
Phenotype
MIM: 103180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARF1 PubMed Links

Gene Details

Gene
ARF1
Gene Name
ADP ribosylation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024226.1 Intron NP_001019397.1
NM_001024227.1 Intron NP_001019398.1
NM_001024228.1 Intron NP_001019399.1
NM_001658.3 Intron NP_001649.1
XM_017001233.1 Intron XP_016856722.1
Gene
C1orf35
Gene Name
chromosome 1 open reading frame 35
There are no transcripts associated with this gene.

Gene
MIR3620
Gene Name
microRNA 3620
There are no transcripts associated with this gene.

View Full Product Details