Product Details
- SNP ID
-
rs1491994
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:190388453 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGATGACTCTCTGGAGGTAAGAAGA[C/T]AGCAGCTTCTTTTCATGATCCAGGC
- Phenotype
-
MIM: 603959
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN16
PubMed Links
Gene Details
- Gene
- CLDN16
- Gene Name
- claudin 16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006580.3 |
|
Intron |
|
|
NP_006571.1 |
View Full Product Details