Product Details

SNP ID
rs1008502
Assay Type
Validated
NCBI dbSNP Submissions
29
Location
Chr.1:185049554 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CCCCTTCAGTGAAGCAACAAGATAT[C/G]TGAGAGCAGTTAGAATTATTGTTTT
Phenotype
MIM: 608985
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF2 PubMed Links

Gene Details

Gene
RNF2
Gene Name
ring finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007212.3 Intron NP_009143.1
XM_005245413.3 Intron XP_005245470.1
XM_011509851.2 Intron XP_011508153.1
XM_011509852.2 Intron XP_011508154.1

View Full Product Details