Product Details

SNP ID
rs784610
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:39660644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTGGGCATGGTCTCAGAGGGGC[A/G]GGGGATCTCAGGGAGGAAGTGAGGG
Phenotype
MIM: 610525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NT5C1A PubMed Links

Gene Details

Gene
NT5C1A
Gene Name
5'-nucleotidase, cytosolic IA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032526.2 Intron NP_115915.1

View Full Product Details