Product Details

SNP ID
rs1044571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96266289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGCGGTACGCAGACGCGCGCGG[C/T]GAGACCCGCTGTCTGCTCAGCGGAC
Phenotype
MIM: 604333 MIM: 601664 MIM: 613403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CIAO1 PubMed Links

Gene Details

Gene
CIAO1
Gene Name
cytosolic iron-sulfur assembly component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004804.2 144 UTR 5 NP_004795.1
Gene
SNRNP200
Gene Name
small nuclear ribonucleoprotein U5 subunit 200
There are no transcripts associated with this gene.

Gene
TMEM127
Gene Name
transmembrane protein 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193304.2 144 Intron NP_001180233.1
NM_017849.3 144 Intron NP_060319.1
XM_017004450.1 144 Intron XP_016859939.1
XM_017004451.1 144 Intron XP_016859940.1
XM_017004452.1 144 Intron XP_016859941.1

View Full Product Details