Product Details

SNP ID
rs1497044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:75291771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTAAGTAATTACAGCTTGCAGTA[A/G]AGTTACTTGGAGTGGTAAATGATTT
Phenotype
MIM: 300656
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UPRT PubMed Links

Gene Details

Gene
UPRT
Gene Name
uracil phosphoribosyltransferase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307944.1 Intron NP_001294873.1
NM_145052.3 Intron NP_659489.1
XM_011530867.2 Intron XP_011529169.1

View Full Product Details