Product Details

SNP ID
rs1062610
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:159028484 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACCAGTGCCTGGTAACCACCTGACA[C/T]GTCCACTCACGCCCACTTCCTCAGC
Phenotype
MIM: 601970
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LINC00689 PubMed Links
Additional Information
For this assay, SNP(s) [rs74392835] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LINC00689
Gene Name
long intergenic non-protein coding RNA 689
There are no transcripts associated with this gene.

Gene
VIPR2
Gene Name
vasoactive intestinal peptide receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304522.1 3716 UTR 3 NP_001291451.1
NM_001308259.1 3716 UTR 3 NP_001295188.1
NM_003382.4 3716 UTR 3 NP_003373.2
XM_005249561.3 3716 UTR 3 XP_005249618.1
XM_006716107.2 3716 Intron XP_006716170.1
XM_006716108.3 3716 Intron XP_006716171.1
XM_011516550.2 3716 Intron XP_011514852.1
XM_017012580.1 3716 UTR 3 XP_016868069.1

View Full Product Details