Product Details

SNP ID
rs1020630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69246543 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACCATTATATAAGGAAGGGATTT[A/T]TACAGGGATTACTAGCCAGTTTTAA
Phenotype
MIM: 604979
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CPSF6 PubMed Links

Gene Details

Gene
CPSF6
Gene Name
cleavage and polyadenylation specific factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300947.1 Intron NP_001287876.1
NM_007007.2 Intron NP_008938.2
XM_005268588.2 Intron XP_005268645.1
XM_005268590.2 Intron XP_005268647.1

View Full Product Details