Product Details
- SNP ID
-
rs1558217
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:11246930 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATTCTCCTTGGTCACATTGCTCAC[G/T]GTCTTCATGTCCCTTCCTGCCCCTC
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79729384] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SHISA6
- Gene Name
- shisa family member 6
View Full Product Details