Product Details

SNP ID
rs1014161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35738676 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTTACAGTTTGGAACCCTGCTT[C/T]CTCCCTTCAATCAAGGGGGAAGGGA
Phenotype
MIM: 611398 MIM: 612128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAS2L2 PubMed Links

Gene Details

Gene
GAS2L2
Gene Name
growth arrest specific 2 like 2
There are no transcripts associated with this gene.

Gene
RASL10B
Gene Name
RAS like family 10 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033315.3 Intron NP_201572.1
XM_017025300.1 Intron XP_016880789.1
XM_017025301.1 Intron XP_016880790.1

View Full Product Details