Product Details

SNP ID
rs712701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127611134 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGAGCCACTAAGACTGGCCAGG[G/T]GACAGTGGGAGGAAGGGCAAGGAAG
Phenotype
MIM: 615800 MIM: 167413
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FSCN3 PubMed Links

Gene Details

Gene
FSCN3
Gene Name
fascin actin-bundling protein 3
There are no transcripts associated with this gene.

Gene
PAX4
Gene Name
paired box 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006193.2 1168 Missense Mutation CAC,CCC H321P NP_006184.2
XM_011516276.2 1168 Missense Mutation CAC,CCC H329P XP_011514578.1

View Full Product Details