Product Details

SNP ID
rs806173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127653848 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATGTGTAATGGAGGGTCAGAACT[C/G]ATCTGAAACTGAATGAGAAGAGAAT
Phenotype
MIM: 602181
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SND1 PubMed Links

Gene Details

Gene
SND1
Gene Name
staphylococcal nuclease and tudor domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017011987.1 Intron XP_016867476.1

View Full Product Details