Product Details
- SNP ID
-
rs806168
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:127665916 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTTAATGAGAAAAGTGGTGCATTCT[A/G]TTCCTCCCGGTGTGTAAGGTAGAGG
- Phenotype
-
MIM: 602181
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SND1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139337816] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SND1
- Gene Name
- staphylococcal nuclease and tudor domain containing 1
View Full Product Details