Product Details

SNP ID
rs1014623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136545335 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTGAGTGTGTACATGTTTGCAAC[G/T]GTGCATATGTATGTGTGTTTGTGTG
Phenotype
MIM: 300583
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR934 PubMed Links

Gene Details

Gene
MIR934
Gene Name
microRNA 934
There are no transcripts associated with this gene.

Gene
VGLL1
Gene Name
vestigial like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016267.3 Intron NP_057351.1

View Full Product Details