Product Details

SNP ID
rs865429
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:43757847 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAAGTGCCAGCTCCCTGGGCAGGCC[A/G]TGGGCAATGCACACTGCAGGGGTGC
Phenotype
MIM: 600183 MIM: 605740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP3 PubMed Links

Gene Details

Gene
DUSP3
Gene Name
dual specificity phosphatase 3
There are no transcripts associated with this gene.

Gene
SOST
Gene Name
sclerostin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025237.2 Intron NP_079513.1

View Full Product Details