Product Details

SNP ID
rs907711
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54498335 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGGAAAAAAAATAGATAATTAG[C/G]TATTTAGATTAAAAAGGGTAGGTGT
Phenotype
MIM: 141180
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
NCKAP1L PubMed Links

Gene Details

Gene
NCKAP1L
Gene Name
NCK associated protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184976.1 Intron NP_001171905.1
NM_005337.4 Intron NP_005328.2

View Full Product Details