Product Details

SNP ID
rs1057838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:6600133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATACCTCCAGCCCCATCATCCT[A/G]AGATAATGAAGCCGTTCATTCTTGG
Phenotype
MIM: 610916
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSUN2 PubMed Links

Gene Details

Gene
NSUN2
Gene Name
NOP2/Sun RNA methyltransferase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193455.1 2478 Silent Mutation CTC,CTT L664L NP_001180384.1
NM_017755.5 2478 Silent Mutation CTC,CTT L699L NP_060225.4

View Full Product Details