Product Details
- SNP ID
-
hCV7570250
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:100205742 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACCTTTTTGGAAAAGAAATGTAT[C/T]AGGTAACAAATTAGAGTATGTATTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTR6
PubMed Links
Gene Details
- Gene
- ACTR6
- Gene Name
- ARP6 actin-related protein 6 homolog
There are no transcripts associated with this gene.
- Gene
- DEPDC4
- Gene Name
- DEP domain containing 4
There are no transcripts associated with this gene.
View Full Product Details