Product Details

SNP ID
rs961697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69887079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATGCCTGGAAAGAGCCCAGATG[A/C]TCATCAGTGGGAGGGCAGATGAGTA
Phenotype
MIM: 608488
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMOC1 PubMed Links

Gene Details

Gene
SMOC1
Gene Name
SPARC related modular calcium binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034852.2 Intron NP_001030024.1
NM_022137.5 Intron NP_071420.1
XM_005267995.1 Intron XP_005268052.1
XM_005267996.1 Intron XP_005268053.1

View Full Product Details