Product Details

SNP ID
rs1475109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111666172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGCTATTCTAATAGCTTTATATG[C/T]GTGAACTCACTTAATTGTTACAGCA
Phenotype
MIM: 604389
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC25-GNG10 PubMed Links

Gene Details

Gene
DNAJC25-GNG10
Gene Name
DNAJC25-GNG10 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004125.3 Intron NP_004116.2
Gene
GNG10
Gene Name
G protein subunit gamma 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017998.3 Intron NP_001017998.1
NM_001198664.1 Intron NP_001185593.1

View Full Product Details