Product Details

SNP ID
rs1056815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:80197713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATTCTCACCACATTTACCCAGG[A/G]CTCTTAGCATCATGTCCCGCATATT
Phenotype
MIM: 601413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIO2 PubMed Links

Gene Details

Gene
DIO2
Gene Name
deiodinase, iodothyronine, type II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000793.5 6087 UTR 3 NP_000784.2
NM_001007023.4 6087 UTR 3 NP_001007024.1
NM_001242502.2 6087 UTR 3 NP_001229431.1
NM_001242503.2 6087 UTR 3 NP_001229432.1
NM_001324462.1 6087 UTR 3 NP_001311391.1
NM_013989.4 6087 UTR 3 NP_054644.1

View Full Product Details