Product Details

SNP ID
rs1329027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:23693920 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGCTTGCTCAAGACCACAGGGCT[A/G]GTAAATTATGGCATTGATACTCAAA
Phenotype
MIM: 601673
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELAVL2 PubMed Links

Gene Details

Gene
ELAVL2
Gene Name
ELAV like neuron-specific RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171195.1 Intron NP_001164666.1
NM_001171197.1 Intron NP_001164668.1
NM_004432.3 Intron NP_004423.2
XM_005251393.3 Intron XP_005251450.1
XM_005251394.3 Intron XP_005251451.1
XM_005251395.3 Intron XP_005251452.1
XM_006716734.3 Intron XP_006716797.1
XM_006716735.3 Intron XP_006716798.1
XM_006716736.3 Intron XP_006716799.1
XM_011517774.2 Intron XP_011516076.1
XM_011517776.2 Intron XP_011516078.1
XM_011517777.2 Intron XP_011516079.1
XM_011517778.2 Intron XP_011516080.2
XM_011517779.2 Intron XP_011516081.1
XM_011517780.2 Intron XP_011516082.1
XM_011517783.2 Intron XP_011516085.1
XM_011517784.2 Intron XP_011516086.1
XM_011517785.2 Intron XP_011516087.1
XM_011517786.2 Intron XP_011516088.1
XM_017014408.1 Intron XP_016869897.1
XM_017014409.1 Intron XP_016869898.1
XM_017014410.1 Intron XP_016869899.1
XM_017014411.1 Intron XP_016869900.1
XM_017014412.1 Intron XP_016869901.1
XM_017014413.1 Intron XP_016869902.1
XM_017014414.1 Intron XP_016869903.1
XM_017014415.1 Intron XP_016869904.1
XM_017014416.1 Intron XP_016869905.1
XM_017014417.1 Intron XP_016869906.1
XM_017014418.1 Intron XP_016869907.1
XM_017014419.1 Intron XP_016869908.1
XM_017014420.1 Intron XP_016869909.1
XM_017014421.1 Intron XP_016869910.1
XM_017014422.1 Intron XP_016869911.1
XM_017014423.1 Intron XP_016869912.1
XM_017014424.1 Intron XP_016869913.1
XM_017014425.1 Intron XP_016869914.1
XM_017014426.1 Intron XP_016869915.1

View Full Product Details