Product Details

SNP ID
rs1005190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:805717 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGCCTGCATTTCTGAGCTCCCAA[C/G]CAGCTCAGGAACAGCGCAGCCGCTC
Phenotype
MIM: 613201 MIM: 607298
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHTF18 PubMed Links

Gene Details

Gene
CHTF18
Gene Name
chromosome transmission fidelity factor 18
There are no transcripts associated with this gene.

Gene
GNG13
Gene Name
G protein subunit gamma 13
There are no transcripts associated with this gene.

Gene
PRR25
Gene Name
proline rich 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013638.1 275 Missense Mutation ACC,AGC T92S NP_001013660.1

View Full Product Details