Product Details
- SNP ID
-
rs1057374
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:121340892 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGTTACAGCCCAGGTTCTTGCCT[C/G]TGGCCTGGGTGCTTAGGGGGTTCAG
- Phenotype
-
MIM: 137350
MIM: 133090
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GSN
PubMed Links
Gene Details
- Gene
- GSN
- Gene Name
- gelsolin
There are no transcripts associated with this gene.
- Gene
- STOM
- Gene Name
- stomatin
View Full Product Details