Product Details

SNP ID
rs1048794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2039927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCCAGAGCCATGCGGCCATCA[A/G]AGACCCTGGGCGGCCGGGCAGAGGG
Phenotype
MIM: 602656 MIM: 606553 MIM: 191092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTHL1 PubMed Links

Gene Details

Gene
NTHL1
Gene Name
nth-like DNA glycosylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318193.1 1046 Silent Mutation CTC,CTT L255L NP_001305122.1
NM_001318194.1 1046 Silent Mutation CTC,CTT L194L NP_001305123.1
NM_002528.6 1046 Silent Mutation CTC,CTT L312L NP_002519.1
XM_017023253.1 1046 Silent Mutation CTG,TTG L341L XP_016878742.1
Gene
SLC9A3R2
Gene Name
SLC9A3 regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130012.2 1046 Intron NP_001123484.1
NM_001252073.1 1046 Intron NP_001239002.1
NM_001252075.1 1046 Intron NP_001239004.1
NM_001252076.1 1046 Intron NP_001239005.1
NM_004785.5 1046 Intron NP_004776.3
XM_005255702.4 1046 Intron XP_005255759.1
XM_006720981.1 1046 Intron XP_006721044.1
XM_017023894.1 1046 Intron XP_016879383.1
XM_017023895.1 1046 Intron XP_016879384.1
Gene
TSC2
Gene Name
tuberous sclerosis 2
There are no transcripts associated with this gene.

View Full Product Details