Product Details

SNP ID
rs700090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:123113262 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACAGCTGTCCAGTGGCTCCCTAC[A/G]GCCCAGAGAATCCAGTCCACACCCC
Phenotype
MIM: 615882 MIM: 611138
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR600 PubMed Links

Gene Details

Gene
MIR600
Gene Name
microRNA 600
There are no transcripts associated with this gene.

Gene
MIR600HG
Gene Name
MIR600 host gene
There are no transcripts associated with this gene.

Gene
RABGAP1
Gene Name
RAB GTPase activating protein 1
There are no transcripts associated with this gene.

Gene
STRBP
Gene Name
spermatid perinuclear RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171137.1 Intron NP_001164608.1
NM_018387.4 Intron NP_060857.2
XM_017014890.1 Intron XP_016870379.1
XM_017014891.1 Intron XP_016870380.1
XM_017014892.1 Intron XP_016870381.1
XM_017014893.1 Intron XP_016870382.1
XM_017014894.1 Intron XP_016870383.1
XM_017014895.1 Intron XP_016870384.1
XM_017014896.1 Intron XP_016870385.1
XM_017014897.1 Intron XP_016870386.1
XM_017014898.1 Intron XP_016870387.1

View Full Product Details