Product Details

SNP ID
rs1294574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90546878 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATTAAGGTCTCATTCCATCCTCT[C/G]TCACCTCCAGGAGCCAGCGCTGCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TTC7B PubMed Links

Gene Details

Gene
TTC7B
Gene Name
tetratricopeptide repeat domain 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010854.1 Intron NP_001010854.1
NM_001320421.1 Intron NP_001307350.1
XM_005267367.1 Intron XP_005267424.1
XM_011536497.1 Intron XP_011534799.1
XM_017021044.1 Intron XP_016876533.1
XM_017021045.1 Intron XP_016876534.1
XM_017021046.1 Intron XP_016876535.1
XM_017021047.1 Intron XP_016876536.1

View Full Product Details