Product Details
- SNP ID
-
rs1035877
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:115842427 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGCCGATCGGGCAGCCGCAGCCCA[G/T]CCGCGCCGCGGAGCCCTGCGCCCAG
- Phenotype
-
MIM: 601507
MIM: 609608
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AP3S1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs371968650,rs542407147] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AP3S1
- Gene Name
- adaptor related protein complex 3 sigma 1 subunit
- Gene
- ATG12
- Gene Name
- autophagy related 12
View Full Product Details