Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001127696.1 | 899 | UTR 3 | NP_001121168.1 | ||
NM_001127697.2 | 899 | UTR 3 | NP_001121169.2 | ||
NM_001164774.1 | 899 | UTR 3 | NP_001158246.1 | ||
NM_001164776.1 | 899 | UTR 3 | NP_001158248.1 | ||
NM_001164777.1 | 899 | UTR 3 | NP_001158249.1 | ||
NM_001164778.1 | 899 | UTR 3 | NP_001158250.1 | ||
NM_001164779.1 | 899 | UTR 3 | NP_001158251.1 | ||
NM_001164780.1 | 899 | UTR 3 | NP_001158252.1 | ||
NM_001164781.1 | 899 | UTR 3 | NP_001158253.1 | ||
NM_001164782.1 | 899 | UTR 3 | NP_001158254.1 | ||
NM_004993.5 | 899 | UTR 3 | NP_004984.2 | ||
NM_030660.4 | 899 | UTR 3 | NP_109376.1 |