Product Details
- SNP ID
-
rs1053932
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:38212849 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CACTCTTGCTATTCTGCATGGGTGA[A/G]GATCGGGTCTTCCAGGAGTTTTCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1755
PubMed Links
Gene Details
- Gene
- KIAA1755
- Gene Name
- KIAA1755
View Full Product Details