Product Details

SNP ID
rs1476703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87885093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGTACCAAGTCCAGAATCTGCA[A/T]ATACTGCAGAAACCACTTCACCTCA
Phenotype
MIM: 604281 MIM: 610821
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DBF4 PubMed Links

Gene Details

Gene
DBF4
Gene Name
DBF4 zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318060.1 838 UTR 5 NP_001304989.1
NM_001318061.1 838 UTR 5 NP_001304990.1
NM_001318062.1 838 UTR 5 NP_001304991.1
NM_006716.3 838 Missense Mutation AAT,TAT N112Y NP_006707.1
Gene
SLC25A40
Gene Name
solute carrier family 25 member 40
There are no transcripts associated with this gene.

View Full Product Details