Product Details

SNP ID
rs8618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74286273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGACTGAACACTGGGAGATTCA[A/G]TGTCCCCCACAGAAACCTAGACCTG
Phenotype
MIM: 603214
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCD4 PubMed Links

Gene Details

Gene
ABCD4
Gene Name
ATP binding cassette subfamily D member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005050.3 2226 UTR 3 NP_005041.1
XM_005267939.3 2226 UTR 3 XP_005267996.1
XM_005267940.3 2226 UTR 3 XP_005267997.1
XM_005267941.4 2226 UTR 3 XP_005267998.1
XM_005267942.4 2226 UTR 3 XP_005267999.1
XM_005267946.3 2226 UTR 3 XP_005268003.1
XM_005267949.3 2226 UTR 3 XP_005268006.1
XM_006720223.2 2226 UTR 3 XP_006720286.1
XM_011537041.2 2226 UTR 3 XP_011535343.1
XM_017021531.1 2226 UTR 3 XP_016877020.1
XM_017021532.1 2226 UTR 3 XP_016877021.1
XM_017021533.1 2226 UTR 3 XP_016877022.1
XM_017021534.1 2226 UTR 3 XP_016877023.1
XM_017021535.1 2226 UTR 3 XP_016877024.1
XM_017021536.1 2226 UTR 3 XP_016877025.1
XM_017021537.1 2226 UTR 3 XP_016877026.1
XM_017021538.1 2226 UTR 3 XP_016877027.1
XM_017021539.1 2226 UTR 3 XP_016877028.1
XM_017021540.1 2226 UTR 3 XP_016877029.1
XM_017021541.1 2226 UTR 3 XP_016877030.1
XM_017021542.1 2226 UTR 3 XP_016877031.1
XM_017021543.1 2226 UTR 3 XP_016877032.1
XM_017021544.1 2226 UTR 3 XP_016877033.1

View Full Product Details