Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001308267.1 | 4761 | UTR 3 | NP_001295196.1 | ||
NM_001308268.1 | 4761 | UTR 3 | NP_001295197.1 | ||
NM_002847.4 | 4761 | UTR 3 | NP_002838.2 | ||
NM_130842.3 | 4761 | UTR 3 | NP_570857.2 | ||
NM_130843.3 | 4761 | UTR 3 | NP_570858.2 | ||
XM_011516446.1 | 4761 | Intron | XP_011514748.1 | ||
XM_011516447.2 | 4761 | Intron | XP_011514749.1 | ||
XM_011516448.2 | 4761 | Intron | XP_011514750.1 | ||
XM_011516449.2 | 4761 | Intron | XP_011514751.1 | ||
XM_017012475.1 | 4761 | UTR 3 | XP_016867964.1 | ||
XM_017012476.1 | 4761 | Intron | XP_016867965.1 |