Product Details

SNP ID
rs1271558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:63315257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACCAGAAGAGCTGAATTTGTCCT[C/G]AACTGTTTTCTCCTACCTTCCCGTA
Phenotype
MIM: 609652
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPHB5 PubMed Links

Gene Details

Gene
GPHB5
Gene Name
glycoprotein hormone beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145171.3 Intron NP_660154.3

View Full Product Details