Product Details

SNP ID
rs1011694
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:81703677 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACTTTCATGAACCATGTAGCCTCCT[A/T]AGAATGCAGTGGCTGGCTTTTGGTA
Phenotype
MIM: 142409
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HGF PubMed Links

Gene Details

Gene
HGF
Gene Name
hepatocyte growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000601.5 Intron NP_000592.3
NM_001010931.2 Intron NP_001010931.1
NM_001010932.2 Intron NP_001010932.1
NM_001010933.2 Intron NP_001010933.1
NM_001010934.2 Intron NP_001010934.1
XM_006715956.2 Intron XP_006716019.1
XM_011516115.2 Intron XP_011514417.1
XM_017012097.1 Intron XP_016867586.1
XM_017012098.1 Intron XP_016867587.1

View Full Product Details