Product Details

SNP ID
rs1254598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52314477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTCTGAGTCTCGGAACGCTCC[A/G]GCTCTCAGACCCTCTTCCTCCCAGG
Phenotype
MIM: 176804
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGER2 PubMed Links

Gene Details

Gene
PTGER2
Gene Name
prostaglandin E receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000956.3 180 UTR 5 NP_000947.2

View Full Product Details