Product Details

SNP ID
rs4644
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:55138217 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGACAGGCACCTCCAGGCGCCTACC[A/C]TGGAGCACCTGGAGCTTATCCCGGA
Phenotype
MIM: 153619
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DLGAP5 PubMed Links

Gene Details

Gene
DLGAP5
Gene Name
DLG associated protein 5
There are no transcripts associated with this gene.

Gene
LGALS3
Gene Name
lectin, galactoside binding soluble 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177388.1 279 Missense Mutation CAT,CCT H64P NP_001170859.1
NM_002306.3 279 Missense Mutation CAT,CCT H64P NP_002297.2
XM_011536759.2 279 Missense Mutation CAT,CCT H78P XP_011535061.2

View Full Product Details