Product Details
- SNP ID
-
rs1055246
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:53944930 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAACCGTGCTGATTTGTTTACTTGG[A/G]TTTGCCAACTGGTCCGATGGATACT
- Phenotype
-
MIM: 602968
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BCAS1
PubMed Links
Gene Details
- Gene
- BCAS1
- Gene Name
- breast carcinoma amplified sequence 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001316361.2 |
1823 |
Missense Mutation |
CCC,TCC |
P505S |
NP_001303290.1 |
NM_001323347.1 |
1823 |
Missense Mutation |
CCC,TCC |
P505S |
NP_001310276.1 |
NM_003657.3 |
1823 |
Missense Mutation |
CCC,TCC |
P583S |
NP_003648.2 |
XM_005260587.2 |
1823 |
Missense Mutation |
CCC,TCC |
P628S |
XP_005260644.1 |
XM_005260589.2 |
1823 |
Missense Mutation |
CCC,TCC |
P614S |
XP_005260646.1 |
XM_005260590.2 |
1823 |
Missense Mutation |
CCC,TCC |
P606S |
XP_005260647.1 |
XM_005260591.2 |
1823 |
Missense Mutation |
CCC,TCC |
P592S |
XP_005260648.1 |
XM_005260593.2 |
1823 |
Missense Mutation |
CCC,TCC |
P569S |
XP_005260650.1 |
XM_005260594.2 |
1823 |
Missense Mutation |
CCC,TCC |
P550S |
XP_005260651.1 |
XM_005260595.2 |
1823 |
Missense Mutation |
CCC,TCC |
P536S |
XP_005260652.1 |
XM_011529090.2 |
1823 |
Missense Mutation |
CCC,TCC |
P628S |
XP_011527392.1 |
XM_011529091.2 |
1823 |
Missense Mutation |
CCC,TCC |
P572S |
XP_011527393.1 |
XM_017028110.1 |
1823 |
Missense Mutation |
CCC,TCC |
P561S |
XP_016883599.1 |
XM_017028111.1 |
1823 |
Missense Mutation |
CCC,TCC |
P547S |
XP_016883600.1 |
XM_017028112.1 |
1823 |
Missense Mutation |
CCC,TCC |
P491S |
XP_016883601.1 |
View Full Product Details