Product Details

SNP ID
rs962961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:56814436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGGTCTCTCACTCCCTGCATTG[C/T]GGTTGCCTCAAAAAAGAGGGTAAGG
Phenotype
MIM: 600037
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTX2 PubMed Links

Gene Details

Gene
OTX2
Gene Name
orthodenticle homeobox 2
There are no transcripts associated with this gene.

Gene
OTX2-AS1
Gene Name
OTX2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details