Product Details

SNP ID
rs1152532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57469361 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCAGCTGTCCTGCTAGGAAGCAG[A/G]AATACCATACAAAGAAGACATGGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf105 PubMed Links

Gene Details

Gene
C14orf105
Gene Name
chromosome 14 open reading frame 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283056.1 3153 UTR 3 NP_001269985.1
NM_001283057.1 3153 UTR 3 NP_001269986.1
NM_001283058.1 3153 UTR 3 NP_001269987.1
NM_001283059.1 3153 UTR 3 NP_001269988.1
NM_001283060.1 3153 Intron NP_001269989.1
NM_018168.3 3153 UTR 3 NP_060638.2
XM_005267806.1 3153 Intron XP_005267863.1
XM_005267810.3 3153 Intron XP_005267867.1
XM_005267811.3 3153 Intron XP_005267868.1
XM_005267813.1 3153 Intron XP_005267870.1
XM_005267814.1 3153 Intron XP_005267871.1
XM_006720188.2 3153 Intron XP_006720251.1
XM_006720189.2 3153 Intron XP_006720252.1

View Full Product Details