Product Details

SNP ID
rs798792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:61570579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAAAGGAGAACCTCCTGAACAG[A/C]ATCCAGTTTGCTTTTAGCTTTAGCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FLJ22447 PubMed Links

Gene Details

Gene
FLJ22447
Gene Name
uncharacterized LOC400221
There are no transcripts associated with this gene.

Gene
LOC101927780
Gene Name
uncharacterized LOC101927780
There are no transcripts associated with this gene.

View Full Product Details