Product Details
- SNP ID
-
rs1049938
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:26190552 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCAGTGGTGGTCTTAGGATCAAAGA[A/C]GACTCATTGGTGTATAGAGTAAGCC
- Phenotype
-
MIM: 600124
MIM: 604135
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HNRNPA2B1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17153378] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HNRNPA2B1
- Gene Name
- heterogeneous nuclear ribonucleoprotein A2/B1
- Gene
- NFE2L3
- Gene Name
- nuclear factor, erythroid 2 like 3
There are no transcripts associated with this gene.
View Full Product Details