Product Details

SNP ID
rs994004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:83475564 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGCAACAATTAGTGTTCTGAGAT[A/G]GAAGCGGTTCCCAGTTTGTAACTTG
Phenotype
MIM: 118661
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VCAN PubMed Links

Gene Details

Gene
VCAN
Gene Name
versican
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126336.2 Intron NP_001119808.1
NM_001164097.1 Intron NP_001157569.1
NM_001164098.1 Intron NP_001157570.1
NM_004385.4 Intron NP_004376.2

View Full Product Details