Product Details
- SNP ID
-
rs1019308
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:12212100 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGCAAGGGATTCCCAGTTTCTCACA[C/G]AGTCTGGCACTTGTCCAAACACCTA
- Phenotype
-
MIM: 613413
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM106B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77287807] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM106B
- Gene Name
- transmembrane protein 106B
View Full Product Details