Product Details

SNP ID
rs3848080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:31852806 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAACCCCACTCAGGACAAAAAAG[A/G]CAAAACACTTGACTCTTTTAAAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EEF1DP3 PubMed Links

Gene Details

Gene
EEF1DP3
Gene Name
eukaryotic translation elongation factor 1 delta pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details