Product Details
- SNP ID
-
rs1800035
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:111078794 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGGGGGCACCGAGCGCAGGCCCAA[C/G]GGTCTGGGCCCCGAGCGCAGCGCGG
- Phenotype
-
MIM: 104210
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADRA2A
PubMed Links
Gene Details
- Gene
- ADRA2A
- Gene Name
- adrenoceptor alpha 2A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000681.3 |
1763 |
Missense Mutation |
AAC,AAG |
N266K |
NP_000672.3 |
View Full Product Details