Product Details

SNP ID
rs1059878
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110900234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCATCAATCTTGGATATTTTT[G/T]TATTTCTATGAATACTATCAATTTT
Phenotype
MIM: 613605 MIM: 608610
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
BBIP1 PubMed Links
Additional Information
For this assay, SNP(s) [rs74378684] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
BBIP1
Gene Name
BBSome interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195304.1 632 UTR 3 NP_001182233.1
NM_001195305.1 632 UTR 3 NP_001182234.1
NM_001195306.1 632 UTR 3 NP_001182235.1
NM_001195307.1 632 UTR 3 NP_001182236.1
NM_001243783.1 632 UTR 3 NP_001230712.1
Gene
MIR4680
Gene Name
microRNA 4680
There are no transcripts associated with this gene.

Gene
PDCD4
Gene Name
programmed cell death 4 (neoplastic transformation inhibitor)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199492.1 632 Intron NP_001186421.1
NM_014456.4 632 Intron NP_055271.2
NM_145341.3 632 Intron NP_663314.1

View Full Product Details