Product Details

SNP ID
rs1060304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:42917334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTTCATCATTCTGCTTGGCAAT[A/G]TAGTCTTCAGAAATCAACTGTGATA
Phenotype
MIM: 176842
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf25 PubMed Links

Gene Details

Gene
C7orf25
Gene Name
chromosome 7 open reading frame 25
There are no transcripts associated with this gene.

Gene
PSMA2
Gene Name
proteasome subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002787.4 993 UTR 3 NP_002778.1

View Full Product Details