Product Details

SNP ID
rs1054972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1852583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGGGTGGGCGCAGGTCTCTCC[A/G]AGCCCCTGGACACTGGAGAGGGGCC
Phenotype
MIM: 606139 MIM: 609614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLF16 PubMed Links

Gene Details

Gene
KLF16
Gene Name
Kruppel like factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031918.3 2703 UTR 3 NP_114124.1
Gene
REXO1
Gene Name
RNA exonuclease 1 homolog
There are no transcripts associated with this gene.

View Full Product Details