Product Details

SNP ID
rs1532046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130344846 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTCTCTCTCTCTTTTACTCTTA[C/T]TCTTTCTCTCTCACTCTCTCTCTTT
Phenotype
MIM: 609561
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPA5 PubMed Links

Gene Details

Gene
CPA5
Gene Name
carboxypeptidase A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127441.1 56 UTR 5 NP_001120913.1
NM_001127442.1 56 UTR 5 NP_001120914.1
NM_001318223.1 56 UTR 5 NP_001305152.1
NM_080385.4 56 UTR 5 NP_525124.3
XM_005250710.1 56 UTR 5 XP_005250767.1
XM_005250712.1 56 UTR 5 XP_005250769.1
XM_011516698.1 56 UTR 5 XP_011515000.1
XM_011516701.1 56 UTR 5 XP_011515003.1
XM_011516702.1 56 UTR 5 XP_011515004.1
XM_011516703.1 56 UTR 5 XP_011515005.1
XM_011516704.1 56 UTR 5 XP_011515006.1
XM_011516705.1 56 UTR 5 XP_011515007.1
Gene
LOC101928632
Gene Name
uncharacterized LOC101928632
There are no transcripts associated with this gene.

View Full Product Details